Please login to your online account to display your discounted pricing

N,N'-Methylenebisacrylamide, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$244.97 - $244.97

Chemical Identifiers

Chemical Name or Material N,N'-Methylenebisacrylamide
CAS 110-26-9
Name Note Purified for Electrophoresis
Purity Grade Ultrapure Bioreagent
Molecular Formula CH2(NHCOCH:CH2)2
Formula Weight 154.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003503

 
avantor performance
403104
Wide Mouth Amber Glass Bottle 100g Each for $244.97
Description & Specifications

Specifications

Formula Weight 154.2
CAS 110-26-9
Chemical Name or Material N,N'-Methylenebisacrylamide
Molecular Formula CH2(NHCOCH:CH2)2
Packaging Wide Mouth Amber Glass Bottle
Purity Grade Ultrapure Bioreagent
Name Note Purified for Electrophoresis
Quantity 100g
Specific Gravity 1L = 1.24kg