missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Methylenebisacrylamide, 99+%

Manufacturer:  Alfa Aesar™ 4370122

 View more versions of this product

Catalog No. AA4370122



Specifications

Specifications

N,N'-Methylenebisacrylamide
White
100g
C7H10N2O2
1706297
It is soluble in water (20g/L) at 20°C,ethanol,and methanol.
C=CC(=O)NCNC(=O)C=C
154.169
154.17
≥99%
1.235 g/mL
>300°C
110-26-9
MFCD00008625
2-propenamide, n,n'-methylenebis, bis-acrylamide, bisacrylamide, methylenebisacrylamide, methylenediacrylamide, n,n'-methylene-bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylidenebisacrylamide
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
N-[(prop-2-enoylamino)methyl]prop-2-enamide
8041
Powder

Chemical Identifiers

110-26-9
154.169
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
8041
C=CC(=O)NCNC(=O)C=C
C7H10N2O2
MFCD00008625
2-propenamide, n,n'-methylenebis, bis-acrylamide, bisacrylamide, methylenebisacrylamide, methylenediacrylamide, n,n'-methylene-bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylidenebisacrylamide
N-[(prop-2-enoylamino)methyl]prop-2-enamide
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 203-750-9

RTECSNumber : AS3678000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents