missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N'-Methylenebisacrylamide, ≥99.9%, Molecular Biology Grade, Ultrapure, Affymetrix/USB™

Manufacturer:  Thermo Fisher Scientific J7582122

 View more versions of this product

Catalog No. AAJ7582122



Description

Description

  • Cross-linking agent used in forming polyacrylamide gels
  • Porosity of gel is determined by chain length and degree of cross-linking
  • Most sequencing gels use ratio of 1 molecule of cross-linker for every 19 monomers of acrylamide
Specifications

Specifications

N,N'-Methylenebisacrylamide
>300°C
110-26-9
(CH2=CHCONH)2CH2
2-propenamide, n,n'-methylenebis
C=CC(=O)NCNC(=O)C=C
154.169
154.17
≥99.9%
Plastic bottle
100g
C7H10N2O2
MFCD00008625
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
N-[(prop-2-enoylamino)methyl]prop-2-enamide
8041
Crystalline Powder
Molecular Biology

Chemical Identifiers

110-26-9
154.169
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
8041
C=CC(=O)NCNC(=O)C=C
C7H10N2O2
MFCD00008625
2-propenamide, n,n'-methylenebis
N-[(prop-2-enoylamino)methyl]prop-2-enamide
Safety and Handling

Safety and Handling

TSCA : Y

Documents