Please login to your online account to display your discounted pricing

N,N'-Methylenebisacrylamide 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  163800050

 View more versions of this product

Catalog No. AC163800050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material N, N'-Methylenebisacrylamide
Color White
Quantity 5g
CAS 110-26-9
Infrared Spectrum Authentic
Packaging Glass bottle
Water 1% Max. (K.F.)
Purity Grade Extra Pure
Physical Form Crystalline Powder
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H10N2O2
Linear Formula (H2C=CHCONH)2CH2
Formula Weight 154.17
Synonym MBA

Specifications

Chemical Name or Material N, N'-Methylenebisacrylamide
CAS 110-26-9
Purity Grade Extra Pure
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H10N2O2
Linear Formula (H2C=CHCONH)2CH2
Formula Weight 154.17
Synonym MBA