Please login to your online account to display your discounted pricing

N,N'-Methylene-bis-acrylamide, Electrophoresis Grade, MP Biomedicals™


Quantity 25g

Manufacturer: mp biomedicals  0219531625

 View more versions of this product

Catalog No. ICN19531625

  • / Each
 

Description & Specifications

Chemical Identifiers

Density 1.235g/mL
Physical State Solid
Quantity 25g
Chemical Name or Material N, N'-Methylene-bis-acrylamide
CAS 110-26-9
Purity Grade Electrophoresis
Physical Form White powder
Molecular Formula C7H10N2O2
Formula Weight 154.2g/mol
Synonym Methylenebisacrylamide
Recommended Storage Store at room temperature.
EINECS Number 203-750-9

Specifications

Chemical Name or Material N, N'-Methylene-bis-acrylamide
CAS 110-26-9
Purity Grade Electrophoresis
Physical Form White powder
Molecular Formula C7H10N2O2
Formula Weight 154.2g/mol
Synonym Methylenebisacrylamide
Accessories

Store at room temperature.