Please login to your online account to display your discounted pricing

N,N'-Methylene-bis-acrylamide, Electrophoresis Grade, MP Biomedicals™

$68.04 - $340.42

Chemical Identifiers

Chemical Name or Material N, N'-Methylene-bis-acrylamide
CAS 110-26-9
Purity Grade Electrophoresis
Physical Form White powder
Molecular Formula C7H10N2O2
Formula Weight 154.2g/mol
Synonym Methylenebisacrylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19531625

 
mp biomedicals
0219531625
25g Each for $68.04

ICN19531683

 
mp biomedicals
0219531683
250g Each for $340.42

ICN19531680

 
mp biomedicals
0219531680
100g Each for $155.88
Description & Specifications

Specifications

Density 1.235g/mL
Physical State Solid
Quantity 25g
Chemical Name or Material N, N'-Methylene-bis-acrylamide
CAS 110-26-9
Purity Grade Electrophoresis
Physical Form White powder
Molecular Formula C7H10N2O2
Formula Weight 154.2g/mol
Synonym Methylenebisacrylamide
Recommended Storage Store at room temperature.
EINECS Number 203-750-9
Safety and Handling

Store at room temperature.