Please login to your online account to display your discounted pricing

N,N'-Methylene-bis-acrylamide, 98% max., Molecular biology reagent grade, MP Biomedicals™


Quantity 250g

Manufacturer: mp biomedicals  0219399783

 View more versions of this product

Catalog No. ICN19399783

  • / Each
 

Description & Specifications

Chemical Identifiers

Density 1.235g/mL
Quantity 250g
Chemical Name or Material N, N'-Methylene-bis-acrylamide
Name Note Nucleic Acid Gel Electrophoresis and Blotting, Protein Electrophoresis and Blotting, Nucleic Acid Electrophoresis
CAS 110-26-9
Purity Grade Molecular Biology Reagent
Physical Form White powder
Molecular Formula C7H10N2O2
Formula Weight 154.2g/mol
MDL Number MFCD00008625
Synonym Methylenebisacrylamide
Recommended Storage Store at room temperature.
EINECS Number 2037509

Specifications

Chemical Name or Material N, N'-Methylene-bis-acrylamide
Name Note Nucleic Acid Gel Electrophoresis and Blotting, Protein Electrophoresis and Blotting, Nucleic Acid Electrophoresis
CAS 110-26-9
Purity Grade Molecular Biology Reagent
Physical Form White powder
Molecular Formula C7H10N2O2
Formula Weight 154.2g/mol
MDL Number MFCD00008625
Synonym Methylenebisacrylamide
Accessories

Store at room temperature.