Please login to your online account to display your discounted pricing

N,N'-Methylene-bis-acrylamide, 98% max., Molecular biology reagent grade, MP Biomedicals™

$160.44 - $344.29

Chemical Identifiers

Chemical Name or Material N, N'-Methylene-bis-acrylamide
Name Note Nucleic Acid Gel Electrophoresis and Blotting, Protein Electrophoresis and Blotting, Nucleic Acid Electrophoresis
CAS 110-26-9
Purity Grade Molecular Biology Reagent
Physical Form White powder
Molecular Formula C7H10N2O2
Formula Weight 154.2g/mol
MDL Number MFCD00008625
Synonym Methylenebisacrylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19399780

 
mp biomedicals
0219399780
100g Each for $160.44

ICN19399783

 
mp biomedicals
0219399783
250g Each for $344.29
Description & Specifications

Specifications

Density 1.235g/mL
Quantity 100g
Chemical Name or Material N, N'-Methylene-bis-acrylamide
Name Note Nucleic Acid Gel Electrophoresis and Blotting, Protein Electrophoresis and Blotting, Nucleic Acid Electrophoresis
CAS 110-26-9
Purity Grade Molecular Biology Reagent
Physical Form White powder
Molecular Formula C7H10N2O2
Formula Weight 154.2g/mol
MDL Number MFCD00008625
Synonym Methylenebisacrylamide
Recommended Storage Store at room temperature.
EINECS Number 2037509
Safety and Handling

Store at room temperature.