Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(n-Hexyl)thiourea, 97%

Chemical Identifiers

Chemical Name or Material N-(n-Hexyl)thiourea
CAS 21071-27-2
Molecular Formula C7H16N2S
Formula Weight 160.28
MDL Number MFCD00041198
Beilstein 1752337
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2168606

 
alfa aesar™
B2168606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.28
Chemical Name or Material N-(n-Hexyl)thiourea
Melting Point 79° to 83°C
Assay 97%
Quantity 5g
Molecular Formula C7H16N2S
Odor Odorless
MDL Number MFCD00041198
CAS 21071-27-2
Beilstein 1752337
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280f-P330-P501a
Wear protective clothing.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P280f-P330-P501a
Wear protective clothing.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.