missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-n-Hexyl-4-nitrobenzenesulfonamide, 97%

$57.05 - $211.27

Chemical Identifiers

CAS 52374-18-2
Molecular Formula C12H18N2O4S
Molecular Weight (g/mol) 286.346
MDL Number MFCD00438183
InChI Key HMKTZTGMSGRPHW-UHFFFAOYSA-N
Synonym 4-nitro-benzenesulfonic acid hexylamide
PubChem CID 296253
IUPAC Name N-hexyl-4-nitrobenzenesulfonamide
SMILES CCCCCCNS(=O)(=O)C1=CC=C(C=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH58403MA View Documents Alfa Aesar™
H58403MA
10mg Each for $57.05
 
AAH58403MF View Documents Alfa Aesar™
H58403MF
50mg Each for $211.27
 
Specifications

Chemical Identifiers

52374-18-2
286.346
HMKTZTGMSGRPHW-UHFFFAOYSA-N
296253
CCCCCCNS(=O)(=O)C1=CC=C(C=C1)[N+](=O)[O-]
C12H18N2O4S
MFCD00438183
4-nitro-benzenesulfonic acid hexylamide
N-hexyl-4-nitrobenzenesulfonamide

Specifications

10mg
C12H18N2O4S
2667220
HMKTZTGMSGRPHW-UHFFFAOYSA-N
N-hexyl-4-nitrobenzenesulfonamide
296253
97%
52374-18-2
MFCD00438183
4-nitro-benzenesulfonic acid hexylamide
CCCCCCNS(=O)(=O)C1=CC=C(C=C1)[N+](=O)[O-]
286.346
286.34
N-n-Hexyl-4-nitrobenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger