missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-n-Hexyl-4-nitrobenzamide, 97%

$220.53 - $703.20

Chemical Identifiers

CAS 89399-21-3
Molecular Formula C13H18N2O3
Molecular Weight (g/mol) 250.298
MDL Number MFCD00229049
InChI Key YIQCYVHPIUGJKJ-UHFFFAOYSA-N
Synonym benzamide, n-hexyl-4-nitro, n-n-hexyl-4-nitrobenzamide, n-hexyl-4-nitro-benzamide, acmc-20llm0, n-hexyl-p-nitrobenzamide
PubChem CID 4080064
IUPAC Name N-hexyl-4-nitrobenzamide
SMILES CCCCCCNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60994MD View Documents Alfa Aesar™
H60994MD
250mg Each for $220.53
 
AAH6099403 View Documents Alfa Aesar™
H6099403
1g Each for $703.20
 
Specifications

Chemical Identifiers

89399-21-3
250.298
YIQCYVHPIUGJKJ-UHFFFAOYSA-N
4080064
CCCCCCNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
C13H18N2O3
MFCD00229049
benzamide, n-hexyl-4-nitro, n-n-hexyl-4-nitrobenzamide, n-hexyl-4-nitro-benzamide, acmc-20llm0, n-hexyl-p-nitrobenzamide
N-hexyl-4-nitrobenzamide

Specifications

250mg
C13H18N2O3
3348542
YIQCYVHPIUGJKJ-UHFFFAOYSA-N
N-hexyl-4-nitrobenzamide
4080064
97%
89399-21-3
MFCD00229049
benzamide, n-hexyl-4-nitro, n-n-hexyl-4-nitrobenzamide, n-hexyl-4-nitro-benzamide, acmc-20llm0, n-hexyl-p-nitrobenzamide
CCCCCCNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
250.298
250.29
N-n-Hexyl-4-nitrobenzamide