Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(n-Hexadecyl)formamide, 97%

$455.04 - $455.04

Chemical Identifiers

Chemical Name or Material N-(n-Hexadecyl)formamide
CAS 53396-33-1
Molecular Formula HC(O)NH(CH2)15CH3
Formula Weight 269.47
MDL Number MFCD00046135
Synonym N-Formyl-n-hexadecylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2040418

  SDS
alfa aesar™
B2040418
50g Each for $455.04

AAB2040409

  SDS
alfa aesar™
B2040409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material N-(n-Hexadecyl)formamide
CAS 53396-33-1
Assay 97%
Quantity 50g
Molecular Formula HC(O)NH(CH2)15CH3
Formula Weight 269.47
MDL Number MFCD00046135
Synonym N-Formyl-n-hexadecylamine