Please login to your online account to display your discounted pricing

Alfa Aesar™ N-n-Heptylmethylamine, 96%

Chemical Identifiers

Chemical Name or Material N-n-Heptylmethylamine
CAS 36343-05-2
Linear Formula CH3(CH2)6NHCH3
Formula Weight 129.25g/mol
MDL Number MFCD00039980
UN Number UN2734
Beilstein 906719
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6579406

 
alfa aesar™
H65794-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6579414

 
alfa aesar™
H65794-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Refractive Index 1.4230
Density 0.768g/mL
Color Colorless
Boiling Point 53 to 58°C (6mmHg)
Quantity 5g
Sensitivity Moisture Sensitive
Chemical Name or Material N-n-Heptylmethylamine
CAS 36343-05-2
Physical Form Liquid
Assay 96%
Linear Formula CH3(CH2)6NHCH3
Formula Weight 129.25g/mol
MDL Number MFCD00039980
UN Number UN2734
Beilstein 906719
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediate
DOT Information Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediate

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.