missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(n-Heptyl)-n-octylamine, 98+%

Manufacturer:  Alfa Aesar™ L1887003

 View more versions of this product

Catalog No. AAL1887003



Specifications

Specifications

N-(n-Heptyl)-n-octylamine
26627-77-0
MFCD03093076
2346655
HAEBNPLWJOGFDQ-UHFFFAOYSA-N
N-heptyloctan-1-amine
12905714
≥98%
1g
C15H33N
UN2735
1-octanamine, n-heptyl
CCCCCCCCNCCCCCCC
227.436
227.44

Chemical Identifiers

26627-77-0
227.436
HAEBNPLWJOGFDQ-UHFFFAOYSA-N
12905714
CCCCCCCCNCCCCCCC
C15H33N
MFCD03093076
1-octanamine, n-heptyl
N-heptyloctan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Documents