Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(n-Heptyl)-n-octylamine, 98+%


Quantity 250mg

Manufacturer: alfa aesar™  L18870MD

 View more versions of this product

Catalog No. AAL18870MD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 227.44
Chemical Name or Material N-(n-Heptyl)-n-octylamine
CAS 26627-77-0
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C15H33N
Health Hazard 1 Danger
Quantity 250mg
MDL Number MFCD03093076
Beilstein 2346655
UN Number UN2735
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Specifications

Chemical Name or Material N-(n-Heptyl)-n-octylamine
CAS 26627-77-0
Molecular Formula C15H33N
Formula Weight 227.44
MDL Number MFCD03093076
UN Number UN2735
Beilstein 2346655
Accessories

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents