missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N′-Ethylenebisoctadecanamide (mixture of fatty acid amides) (consists of C14, C16 and C18), TCI America™

Manufacturer:  TCI America E024325G

 View more versions of this product

Catalog No. E024325G



Specifications

Specifications

N,N′-Ethylenebisoctadecanamide (mixture of fatty acid amides) (consists of C14, C16 and C18)
144°C
110-30-5
MFCD00059224
RKISUIUJZGSLEV-UHFFFAOYSA-N
N-[2-(octadecanoylamino)ethyl]octadecanamide
8044
Crystal-Powder at 20°C
White-Yellow
25g
C38H76N2O2
N,N′C-Ethylenebisstearamide
CCCCCCCCCCCCCCCCCC(=O)NCCNC(=O)CCCCCCCCCCCCCCCCC
593.038
593.04
≥85.0% (GC)

Chemical Identifiers

110-30-5
593.038
RKISUIUJZGSLEV-UHFFFAOYSA-N
8044
CCCCCCCCCCCCCCCCCC(=O)NCCNC(=O)CCCCCCCCCCCCCCCCC
C38H76N2O2
MFCD00059224
N,N′C-Ethylenebisstearamide
N-[2-(octadecanoylamino)ethyl]octadecanamide
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0831

TSCA : Yes

Documents