Please login to your online account to display your discounted pricing

N,N'-Ethylenebisbenzamide, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC365860050

 View more versions of this product

Catalog No. AC365860050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 268.32
Quantity 5g
Color White
Melting Point 240° to 244°C
CAS Max % 100.0
Chemical Name or Material N, N'-Ethylenebisbenzamide
Assay Percent Range 98%
Molecular Formula C16H16N2O2
CAS 644-33-7
CAS Min % 97.5
Name Note 98%
MDL Number MFCD00267629
Infrared Spectrum Authentic

Specifications

Chemical Name or Material N, N'-Ethylenebisbenzamide
Name Note 98%
CAS 644-33-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H16N2O2
Formula Weight 268.32
MDL Number MFCD00267629