Please login to your online account to display your discounted pricing

N,N'-Ethylenebisbenzamide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N, N'-Ethylenebisbenzamide
Name Note 98%
CAS 644-33-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H16N2O2
Formula Weight 268.32
MDL Number MFCD00267629
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC365860050

  SDS
thermo fisher scientific
AC365860050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 268.32
Quantity 5g
Color White
Melting Point 240° to 244°C
CAS Max % 100.0
Chemical Name or Material N, N'-Ethylenebisbenzamide
Assay Percent Range 98%
Molecular Formula C16H16N2O2
CAS 644-33-7
CAS Min % 97.5
Name Note 98%
MDL Number MFCD00267629
Infrared Spectrum Authentic