missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N'-Diphenylurea, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC161850250

 View more versions of this product

Catalog No. AC161850250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N, N'-Diphenylurea
Glass bottle
White
25g
97%
C6H5NHCONHC6H5
GWEHVDNNLFDJLR-UHFFFAOYSA-N
1,3-diphenylurea
7595
212.25
97%
Authentic
260.0°C to 262.0°C
232.0°C to 238.0°C
102-07-8
C13H12N2O
n,n'-diphenylurea, carbanilide, diphenylurea, diphenylcarbamide, s-diphenylurea, urea, n,n'-diphenyl, sym-diphenylurea, acardite i, acardite, n-phenyl-n'-phenylurea
C1=CC=C(C=C1)NC(=O)NC2=CC=CC=C2
212.252
CHEBI:41320
Crystalline Powder

Chemical Identifiers

102-07-8
212.252
n,n'-diphenylurea, carbanilide, diphenylurea, diphenylcarbamide, s-diphenylurea, urea, n,n'-diphenyl, sym-diphenylurea, acardite i, acardite, n-phenyl-n'-phenylurea
CHEBI:41320
C1=CC=C(C=C1)NC(=O)NC2=CC=CC=C2
C13H12N2O
GWEHVDNNLFDJLR-UHFFFAOYSA-N
7595
1,3-diphenylurea
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents