missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N'-Diphenylformamidine, 98%, ACROS Organics™

Manufacturer:  Acros Organics 170471000

 View more versions of this product

Catalog No. AC170471000


 

Specifications

Specifications

N, N';-Diphenylformamidine
622-15-1
100.0
Glass bottle
98%
HC(=NC6H5)NHC6H5
12, 236
ZQUVDXMUKIVNOW-UHFFFAOYSA-N
C1=CC=C(C=C1)NC=NC2=CC=CC=C2
Undesignated
100g
196.25
98%
97.5
Authentic
≥97.5%
C13H12N2
MFCD00003026
01,339
136°C to 140°C
N,N'-diphenylmethanimidamide
196.253
94888
Crystalline Powder or Crystals

Chemical Identifiers

622-15-1
196.253
ZQUVDXMUKIVNOW-UHFFFAOYSA-N
C13H12N2
MFCD00003026
94888
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 210-720-9

RTECSNumber : PB7705000

TSCA : TSCA

SDS Documents