Please login to your online account to display your discounted pricing

N,N'-Diphenylformamidine, 98%, ACROS Organics™

$44.15 - $117.53

Chemical Identifiers

Chemical Name or Material N, N';-Diphenylformamidine
Name Note 98%
CAS 622-15-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H12N2
Linear Formula HC(=NC6H5)NHC6H5
Formula Weight 196.25
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170470250

  SDS
thermo fisher scientific
170470250
Glass bottle 25g Each for $44.15

AC170471000

  SDS
thermo fisher scientific
170471000
Glass bottle 100g Each for $117.53
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 196.25
Physical Form Crystalline Powder or Crystals
Quantity 25g
Color White to yellow to beige
TSCA TSCA
Melting Point 136° to 140°C
RTECS Number PB7705000
EINECS Number 210-720-9
Beilstein 12, 236
Fieser 01,339
CAS Max % 100.0
Packaging Glass bottle
CAS Min % 97.5
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 1 MOISTURE SENSITIVE
Name Note 98%
Assay Percent Range 98%
Linear Formula HC(=NC6H5)NHC6H5
MDL Number MFCD00003026
Chemical Name or Material N, N';-Diphenylformamidine
Molecular Formula C13H12N2
CAS 622-15-1
Safety and Handling

MOISTURE SENSITIVE

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.