Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Diphenylformamidine, 98%

$102.68 - $102.68

Chemical Identifiers

Chemical Name or Material N,N'-Diphenylformamidine
CAS 622-15-1
Molecular Formula C13H12N2
Formula Weight 196.25
MDL Number MFCD00003026
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2378622

 
alfa aesar™
B2378622
100g Each for $102.68
Description & Specifications

Specifications

Formula Weight 196.25
Chemical Name or Material N,N'-Diphenylformamidine
TSCA TSCA
Melting Point 136° to 139°C
Assay 98%
Quantity 100g
Molecular Formula C13H12N2
MDL Number MFCD00003026
CAS 622-15-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
RTECS Number PB7705000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C