missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N'-Diphenylbenzidine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 117210050

 View more versions of this product

Catalog No. AC117210050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

N, N'-Diphenylbenzidine
96.0
246°C to 250°C
Authentic
Glass bottle
MFCD00003016
13, 223
n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine
N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1
336.44
336.42
Fine Crystalline Powder
531-91-9
100.0
Brown to Gray
96% min. (HPLC)
C24H20N2
5 g
15, 3349
FDRNXKXKFNHNCA-UHFFFAOYSA-N
4-(4-anilinophenyl)-N-phenylaniline
68280
97%

Chemical Identifiers

531-91-9
336.44
FDRNXKXKFNHNCA-UHFFFAOYSA-N
68280
N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1
C24H20N2
MFCD00003016
n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine
4-(4-anilinophenyl)-N-phenylaniline
Safety and Handling

Safety and Handling

EINECSNumber : 208-521-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only