Please login to your online account to display your discounted pricing

N,N'-Dimethylthiourea, 99%, ACROS Organics™

$59.98 - $59.98

Chemical Identifiers

Chemical Name or Material N, N'-Dimethylthiourea
Name Note 99%
CAS 534-13-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C3H8N2S
Linear Formula CH3NHCSNHCH3
Formula Weight 104.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC116891000

  SDS
fisher scientific acros
116891000
Glass bottle 100g Each for $59.98

AC116890050

  SDS
thermo fisher scientific
116890050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 104.18
Physical Form Crystals, Crystalline Powder and/or Chunks
Quantity 100g
Color Colorless to white
Merck Index 15, 3287
Beilstein 04, 70
Melting Point 61° to 67°C
RTECS Number YT1520000
TSCA TSCA
EINECS Number 208-588-2
CAS Max % 100.0
Packaging Glass bottle
Synonym Dimethylthiocarbamide
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 1 Warning
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula CH3NHCSNHCH3
Molecular Formula C3H8N2S
Assay Percent Range 99%
Chemical Name or Material N, N'-Dimethylthiourea
CAS 534-13-4
Name Note 99%
MDL Number MFCD00004923
Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.