Learn More
N,N-Dimethylthioformamide, 97%, Thermo Scientific Chemicals
$79.23 - $899.89
Chemical Identifiers
CAS | 758-16-7 |
---|---|
Molecular Formula | C3H7NS |
Molecular Weight (g/mol) | 89.156 |
MDL Number | MFCD00004943 |
InChI Key | SKECXRFZFFAANN-UHFFFAOYSA-N |
Synonym | n,n-dimethylthioformamide, dimethylthioformamide, methanethioamide, n,n-dimethyl, formamide, n,n-dimethylthio, nn-dimethylthioformamide, methanethioamide,n,n-dimethyl, formamide,n-dimethylthio, dimethylamino methanethial, n,n-dimethyl-thioformamide, methanethioamide, dimethyl |
PubChem CID | 69794 |
IUPAC Name | N,N-dimethylmethanethioamide |
SMILES | CN(C)C=S |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2452906
|
Thermo Scientific Chemicals
B2452906 |
5 g |
Each for $79.23
|
|
|||||
AAB2452914
|
Thermo Scientific Chemicals
B2452914 |
25 g |
Each for $313.71
|
|
|||||
AAB2452922
|
Thermo Scientific Chemicals
B2452922 |
100 g |
Each for $899.89
|
|
|||||
Description
It is used in telecommunication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in telecommunication.
Solubility
Fully miscible in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Chemical Identifiers
758-16-7 | |
89.156 | |
SKECXRFZFFAANN-UHFFFAOYSA-N | |
69794 | |
CN(C)C=S |
C3H7NS | |
MFCD00004943 | |
n,n-dimethylthioformamide, dimethylthioformamide, methanethioamide, n,n-dimethyl, formamide, n,n-dimethylthio, nn-dimethylthioformamide, methanethioamide,n,n-dimethyl, formamide,n-dimethylthio, dimethylamino methanethial, n,n-dimethyl-thioformamide, methanethioamide, dimethyl | |
N,N-dimethylmethanethioamide |
Specifications
758-16-7 | |
C3H7NS | |
5 g | |
Fully miscible in water. | |
CN(C)C=S | |
89.156 | |
89.15 | |
N,N-Dimethylthioformamide |
58°C to 60°C (1 mmHg) | |
MFCD00004943 | |
n,n-dimethylthioformamide, dimethylthioformamide, methanethioamide, n,n-dimethyl, formamide, n,n-dimethylthio, nn-dimethylthioformamide, methanethioamide,n,n-dimethyl, formamide,n-dimethylthio, dimethylamino methanethial, n,n-dimethyl-thioformamide, methanethioamide, dimethyl | |
SKECXRFZFFAANN-UHFFFAOYSA-N | |
N,N-dimethylmethanethioamide | |
69794 | |
97% |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
H312+H332
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 212-060-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only