missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethylthioacetamide, 97%

$54.00 - $195.96

Chemical Identifiers

CAS 631-67-4
Molecular Formula C4H9NS
Formula Weight 103.19
MDL Number MFCD00022178
Chemical Name or Material N,N-Dimethylthioacetamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2002003 SDS Alfa Aesar™
B2002003
1g Each for $54.00
AAB2002006 SDS Alfa Aesar™
B2002006
5g Each for $195.96
Specifications

Description

Applications
N,N-Dimethylthioacetamide is used as pharmaceutical intermediates.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Description & Specifications

Chemical Identifiers

631-67-4
103.19
N,N-Dimethylthioacetamide
C4H9NS
MFCD00022178

Specifications

631-67-4
125° to 127°C (22mmHg)
C4H9NS
MFCD00022178
97%
72° to 75°C
1g
103.19
N,N-Dimethylthioacetamide

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA