missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethylpropionamide, ≥98%

Manufacturer:  Alfa Aesar™ H6485036

 View more versions of this product

Catalog No. AAH6485036



Description

Description

Applications
N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.

Solubility
Soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Specifications

N,N-Dimethylpropionamide
1.4400
Colorless
500g
C5H11NO
MFCD00009301
n,n-dimethylpropionamide, propanamide, n,n-dimethyl, dimethylamide of propionic acid, propionamide, n,n-dimethyl, propionamide, n,n-dimethyl, n,n-dimethyl propionamide, acmc-209p0o, propionic acid dimethylamide, 4-04-00-00184 beilstein handbook reference, ksc494e1j
CCC(=O)N(C)C
101.149
101.15
≥98%
0.920g/mL
174°C to 175°C (literature)
-45°C (literature)
758-96-3
CH3CH2CON(CH3)2
1740719
MBHINSULENHCMF-UHFFFAOYSA-N
N,N-dimethylpropanamide
12965
Liquid

Chemical Identifiers

758-96-3
101.149
MBHINSULENHCMF-UHFFFAOYSA-N
12965
CCC(=O)N(C)C
C5H11NO
MFCD00009301
n,n-dimethylpropionamide, propanamide, n,n-dimethyl, dimethylamide of propionic acid, propionamide, n,n-dimethyl, propionamide, n,n-dimethyl, n,n-dimethyl propionamide, acmc-209p0o, propionic acid dimethylamide, 4-04-00-00184 beilstein handbook reference, ksc494e1j
N,N-dimethylpropanamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UE3810000

TSCA : TSCA

Documents