Learn More
N,N-Dimethylpropionamide, ≥98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6485014
Description
N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.
Solubility
Soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
N,N-Dimethylpropionamide | |
-45°C (literature) | |
0.920 g/mL | |
62°C (144°F) | |
1.44 | |
MFCD00009301 | |
1740719 | |
Soluble in water. | |
CCC(=O)N(C)C | |
101.149 | |
101.15 | |
Liquid |
758-96-3 | |
Colorless | |
174°C to 175°C (literature) | |
C5H11NO | |
CH3CH2CON(CH3)2 | |
25 g | |
n,n-dimethylpropionamide, propanamide, n,n-dimethyl, dimethylamide of propionic acid, propionamide, n,n-dimethyl, n,n-dimethyl propionamide, acmc-209p0o, propionic acid dimethylamide, 4-04-00-00184 beilstein handbook reference, ksc494e1j | |
MBHINSULENHCMF-UHFFFAOYSA-N | |
N,N-dimethylpropanamide | |
12965 | |
≥98% |
Chemical Identifiers
758-96-3 | |
101.149 | |
MBHINSULENHCMF-UHFFFAOYSA-N | |
12965 | |
CCC(=O)N(C)C |
C5H11NO | |
MFCD00009301 | |
n,n-dimethylpropionamide, propanamide, n,n-dimethyl, dimethylamide of propionic acid, propionamide, n,n-dimethyl, n,n-dimethyl propionamide, acmc-209p0o, propionic acid dimethylamide, 4-04-00-00184 beilstein handbook reference, ksc494e1j | |
N,N-dimethylpropanamide |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 212-064-9
RTECSNumber : UE3810000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only