missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylphosphoramidodichloridate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0723106

 View more versions of this product

Catalog No. AAL0723106



Description

Description

N,N-Dimethylphosphoramidodichloridate is used in the synthesis of phosphoramidates as well as (RS)-ethyl N,N-dimethylphosphoramidocyanidate (Nerve agent GA) which is used as a chemical weapon. It forms active intermediates, which react with alcohols in situ to give esters. It is also used to convert primary alcohols to alkyl chlorides. Furthe, it serves as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. In addition to this, it reacts with 1,2 diols to prepare cyclic phosphoric amides followed by alkenes after reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylphosphoramidodichloridate is used in the synthesis of phosphoramidates as well as (RS)-ethyl N,N-dimethylphosphoramidocyanidate (Nerve agent GA) which is used as a chemical weapon. It forms active intermediates, which react with alcohols in situ to give esters. It is also used to convert primary alcohols to alkyl chlorides. Furthe, it serves as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. In addition to this, it reacts with 1,2 diols to prepare cyclic phosphoric amides followed by alkenes after reduction.

Solubility
Miscible with dimethoxyethane.

Notes
Moisture and air sensitive. Hygroscopic. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

N,N-Dimethylphosphoramidodichloridate
1.36
>110°C (230°F)
1.463
5 g
969956
dimethylphosphoramidic dichloride, n,n-dimethylphosphoramidodichloridate, dichlorophosphoric dimethylamide, phosphoramidic dichloride, dimethyl, dimethylamido phosphoric dichloride, dimethylamidophosphoric acid dichloride, dimethylaminophosphoryl dichloride, n,n-dimethylphosphoramidic dichloride, dimethylamidophosphoric dichloride, n,n-dimethylphosphoramic dichloride
YNHXBEVSSILHPI-UHFFFAOYSA-N
N-dichlorophosphoryl-N-methylmethanamine
12673
97%
677-43-0
89°C to 90°C (20 mmHg)
C2H6Cl2NOP
MFCD00013896
UN3265
Moisture sensitive
Miscible with dimethoxyethane.
CN(C)P(=O)(Cl)Cl
161.95
161.96

Chemical Identifiers

677-43-0
161.95
YNHXBEVSSILHPI-UHFFFAOYSA-N
12673
CN(C)P(=O)(Cl)Cl
C2H6Cl2NOP
MFCD00013896
dimethylphosphoramidic dichloride, n,n-dimethylphosphoramidodichloridate, dichlorophosphoric dimethylamide, phosphoramidic dichloride, dimethyl, dimethylamido phosphoric dichloride, dimethylamidophosphoric acid dichloride, dimethylaminophosphoryl dichloride, n,n-dimethylphosphoramidic dichloride, dimethylamidophosphoric dichloride, n,n-dimethylphosphoramic dichloride
N-dichlorophosphoryl-N-methylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 211-641-2

RTECSNumber : TB4025000

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only