missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Dimethyloxamide, 98%

Manufacturer:  Alfa Aesar™ A1133314

 View more versions of this product

Catalog No. AAA1133314



Specifications

Specifications

N,N'-Dimethyloxamide
25g
C4H8N2O2
1753961
IPZCJUOJSODZNK-UHFFFAOYSA-N
N,N'-dimethyloxamide
69204
98%
215°C to 217°C
615-35-0
MFCD00025634
n,n'-dimethyloxalamide, nn'-dimethyloxamide, n1,n2-dimethyloxalamide, ethanediamide, n,n'-dimethyl, unii-573d5i9hqv, 4-oxalic acid n-methyl amide, ch3nhcoconhch3, n,n'-dimethylethanediamide, n,n/'-dimethyloxamide, acmc-1bg8g
CNC(=O)C(=O)NC
116.12
116.12

Chemical Identifiers

615-35-0
116.12
IPZCJUOJSODZNK-UHFFFAOYSA-N
69204
CNC(=O)C(=O)NC
C4H8N2O2
MFCD00025634
n,n'-dimethyloxalamide, nn'-dimethyloxamide, n1,n2-dimethyloxalamide, ethanediamide, n,n'-dimethyl, unii-573d5i9hqv, 4-oxalic acid n-methyl amide, ch3nhcoconhch3, n,n'-dimethylethanediamide, n,n/'-dimethyloxamide, acmc-1bg8g
N,N'-dimethyloxamide
SDS Documents