missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethylisobutyramide, 99%

Manufacturer:  Alfa Aesar™ H5343022

 View more versions of this product

Catalog No. AAH5343022



Description

Description

Applications
1- chloro-N,N-2-trimethylpropenylamine readily prepared′ from N,N-dimethylisobutyramide. Isoxazoles are prepared by deprotonation of N,N-dimethylisobutyramide.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

N,N-Dimethylisobutyramide
154°C (309°F)
175°C to 176°C (44mmHg)
21678-37-5
(CH3)2CHCON(CH3)2
1742178
Soluble in water.
CC(C)C(=O)N(C)C
115.176
115.18
0.892
1.439
100g
C6H13NO
MFCD00075378
acmc-20ak9x
GXMIHVHJTLPVKL-UHFFFAOYSA-N
N,N,2-trimethylpropanamide
243415
99%

Chemical Identifiers

21678-37-5
115.176
GXMIHVHJTLPVKL-UHFFFAOYSA-N
243415
CC(C)C(=O)N(C)C
C6H13NO
MFCD00075378
acmc-20ak9x
N,N,2-trimethylpropanamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents