missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylformamide, Spectrophotometric Grade, >99.7%, Alfa Aesar™

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%

Manufacturer:  Alfa Aesar 13808K7

 View more versions of this product

Catalog No. AA13808K7



Specifications

Specifications

Colorless liquid
0.944g/cm3
Amine-like
Colorless
Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%
>99.7%
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
N,N-dimethylformamide
6228
73.09
Sprectrophotometric
N, N-Dimethylformamide
440°C (824°F)
3.5 hPa (3mm Hg) at 20°C
4L
68-12-2
C3H7NO
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CN(C)C=O
73.095
CHEBI:17741
Liquid

Chemical Identifiers

68-12-2
73.095
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
N,N-dimethylformamide
C3H7NO
MFCD00003284
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CHEBI:17741
CN(C)C=O
Safety and Handling

Safety and Handling

H360-H226-H312-H332-H319: May damage fertility or the unborn child. Flammable liquid and vapour. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation.

DOTInformation : Hazard Class: 3, Packing Group: III

EINECSNumber : 200-679-5

Documents