Please login to your online account to display your discounted pricing

N,N-Dimethylformamide, For Spectroscopy Uvasol™, EMD Millipore

For spectroscopy Uvasol™

$223.89 - $839.09

Chemical Identifiers

Chemical Name or Material N,N-Dimethylformamide
CAS 68-12-2
Molecular Formula HCON(CH3)2
Formula Weight 73.09
UN Number 2265
Synonym DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1029370500

 
emd millipore corporation
1029370500
Glass Bottle 500mL Each for $223.89

M1029372500

 
emd millipore corporation
1029372500
Glass Bottle 2.5L Each for $839.09
Description & Specifications

Specifications

Boiling Point 153°C
Melting Point −61°C
pH 7 (200g/L at 20°C)
Formula Weight 73.09
Quantity 500mL
Storage Information 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.
CAS 68-12-2
Synonym DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide
Health Hazard 1 H360D May damage the unborn child.
Health Hazard 2 H226 Flammable liquid and vapor.
Chemical Name or Material N,N-Dimethylformamide
Ignition Point 410°C (770°F)
Odor Weakly amine-like
Packaging Glass Bottle
Refractive Index 1.43
Vapor Pressure 3.77 hPa (20°C)
Flash Point 58°C (136°F)
Physical Form Liquid
Color Colorless
Molecular Formula HCON(CH3)2
UN Number 2265
Solubility Information Soluble in water at 20°C
Description The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The Uvasol™ solvents offer best UV transmittance.
Density 0.94g/cm3 at 20°C
Health Hazard 3 H312 + H332 Harmful in contact with skin or if inhaled.
Safety and Handling

H360D May damage the unborn child.

H312 + H332 Harmful in contact with skin or if inhaled.

H226 Flammable liquid and vapor.