Please login to your online account to display your discounted pricing

N,N-Dimethylformamide, EMD Millipore


For headspace gas chromatography SupraSolv™

Quantity 500mL
Packaging Glass Bottle

Manufacturer: emd millipore corporation  1002020500

 View more versions of this product

Catalog No. M1002020500

  • / Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 153°C
Melting Point −61°C
pH 7 (200g/L at 20°C)
Formula Weight 73.09
Quantity 500mL
Description SupraSolv™ solvents for headspace gas chromatography are developed particularly for the analysis of residual solvents in drug substances, excipients and drug products according to ICH, Ph Eur and USP - SupraSolv headspace solvents are tested for all residual solvents defined by the ICH.
Storage Information 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.
Odor Weakly amine-like
UN Number 2265
Health Hazard 3 H312 + H332 Harmful in contact with skin or if inhaled.
Physical Form Liquid
Color Colorless
Molecular Formula HCON(CH3)2
Synonym DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide
Ignition Point 410°C (770°F)
Packaging Glass Bottle
Health Hazard 1 H360D May damage the unborn child.
CAS 68-12-2
Health Hazard 2 H226 Flammable liquid and vapor.
Density 0.94g/cm3 at 20°C
Refractive Index 1.43
Vapor Pressure 3.77 hPa (20°C)
Flash Point 58°C (136°F)
Solubility Information Soluble in water at 20°C
Chemical Name or Material N,N-Dimethylformamide

Specifications

Chemical Name or Material N,N-Dimethylformamide
CAS 68-12-2
Molecular Formula HCON(CH3)2
Formula Weight 73.09
UN Number 2265
Synonym DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide
Accessories

H360D May damage the unborn child.

H312 + H332 Harmful in contact with skin or if inhaled.

H226 Flammable liquid and vapor.