missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Dimethylformamide, MilliporeSigma™
For headspace gas chromatography SupraSolv™
$311.89 - $1,153.40
Chemical Identifiers
CAS | 68-12-2 |
---|---|
Molecular Formula | C3H7NO |
Molecular Weight (g/mol) | 73.095 |
InChI Key | ZMXDDKWLCZADIW-UHFFFAOYSA-N |
Synonym | dimethyl formamide, dimethylformamid, dimethylformamide, dimetilformamide, dmf, dmfa, dwumetyloformamid, formamide, n,n-dimethyl, n,n-dimethylmethanamide, n-formyldimethylamine |
PubChem CID | 6228 |
ChEBI | CHEBI:17741 |
IUPAC Name | N,N-dimethylformamide |
SMILES | CN(C)C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
M1002020500
|
Emd Millipore Corporation
1002020500 |
500mL | Glass Bottle |
Each for $311.89
|
|
36-103-1321
|
Emd Millipore Corporation
1002021000 |
1L | Glass Bottle |
Each for $556.06
|
|
M1002022500
|
Emd Millipore Corporation
1002022500 |
2.5L | Glass Bottle |
Each for $1,153.40
|
|
Chemical Identifiers
68-12-2 | |
73.095 | |
dimethyl formamide, dimethylformamid, dimethylformamide, dimetilformamide, dmf, dmfa, dwumetyloformamid, formamide, n,n-dimethyl, n,n-dimethylmethanamide, n-formyldimethylamine | |
CHEBI:17741 | |
CN(C)C=O |
C3H7NO | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
Specifications
68-12-2 | |
2265 | |
Soluble in water at 20°C | |
CN(C)C=O | |
73.095 | |
CHEBI:17741 | |
Liquid | |
0.94g/cm3 at 20°C | |
Weakly amine-like | |
1.43 | |
3.77 hPa (20°C) | |
153°C | |
-61°C | |
500mL |
C3H7NO | |
dimethyl formamide, dimethylformamid, dimethylformamide, dimetilformamide, dmf, dmfa, dwumetyloformamid, formamide, n,n-dimethyl, n,n-dimethylmethanamide, n-formyldimethylamine | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.09 | |
SupraSolv™ solvents for headspace gas chromatography are developed particularly for the analysis of residual solvents in drug substances, excipients and drug products according to ICH, Ph Eur and USP - SupraSolv headspace solvents are tested for all residual solvents defined by the ICH. | |
410°C (770°F) | |
Glass Bottle | |
5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons. | |
58°C (136°F) | |
Colorless | |
7 (at 200g/L 20°C) | |
N,N-Dimethylformamide |
Safety and Handling
H226 Flammable liquid and vapor.
H312 + H332 Harmful in contact with skin or if inhaled.
H360D May damage the unborn child.