Please login to your online account to display your discounted pricing

N,N-Dimethylformamide dipropyl acetal, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  176480050

 View more versions of this product

Catalog No. AC176480050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 175.27
Physical Form Liquid
Quantity 5g
Color Clear colorless to light yellow
EINECS Number 227-855-4
Density 0.85g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4080 to 1.4100 (20°C, 589nm)
Synonym 1, 1-Dipropoxytrimethylamine
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material N, N-Dimethylformamide dipropyl acetal
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H21NO2
Linear Formula (CH3)2NCH(OCH2CH2CH3)2
MDL Number MFCD00009374
CAS 6006-65-1
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.

Specifications

Chemical Name or Material N, N-Dimethylformamide dipropyl acetal
Name Note 98%
CAS 6006-65-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H21NO2
Linear Formula (CH3)2NCH(OCH2CH2CH3)2
Formula Weight 175.27
MDL Number MFCD00009374
Synonym 1, 1-Dipropoxytrimethylamine
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents