Please login to your online account to display your discounted pricing

N,N-Dimethylformamide dipropyl acetal, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N, N-Dimethylformamide dipropyl acetal
Name Note 98%
CAS 6006-65-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H21NO2
Linear Formula (CH3)2NCH(OCH2CH2CH3)2
Formula Weight 175.27
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC176480050

  SDS
thermo fisher scientific
176480050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 175.27
Physical Form Liquid
Quantity 5g
Color Clear colorless to light yellow
EINECS Number 227-855-4
Density 0.85g/mL
CAS Max % 100.0
Refractive Index 1.4080 to 1.4100 (20°C, 589nm)
Synonym 1, 1-Dipropoxytrimethylamine
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
MDL Number MFCD00009374
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Name Note 98%
Chemical Name or Material N, N-Dimethylformamide dipropyl acetal
Assay Percent Range 98%
Linear Formula (CH3)2NCH(OCH2CH2CH3)2
Molecular Formula C9H21NO2
CAS 6006-65-1

Alkylation reagent

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.