Learn More
Alfa Aesar™ N,N-Dimethylformamide dimethyl acetal, 97%
Manufacturer: Alfa Aesar™ A1535014
Description
ApplicationsN,N-Dimethylformamide dimethyl acetal is used as an intermediate in the formation of pyridine derivatives, which exhibits inhibition against PI3 kinase p110alfa enzymes. It is utilized for the derivatization of primary sulfonamides and trifluoroacetic acid. It is also used in the preparation of formamidine derivatives. It is used as a reagent for n-dimethylaminomethylene and methyl esters. Further, it is used to catalyze the coupling of epoxides with carbon dioxide to prepare cyclic carbonates.
Solubility
Miscible with most organic solvents.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.
Specifications
N,N-Dimethylformamide dimethyl acetal | |
102°C to 104°C | |
25g | |
C5H13NO2 | |
UN1993 | |
ZSXGLVDWWRXATF-UHFFFAOYSA-N | |
1,1-dimethoxy-N,N-dimethylmethanamine | |
78373 | |
119.16 |
7°C (45°F) | |
-85°C | |
4637-24-5 | |
MFCD00008482 | |
n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane | |
CN(C)C(OC)OC | |
119.164 | |
CHEBI:85061 | |
97% |
Chemical Identifiers
4637-24-5 | |
119.164 | |
ZSXGLVDWWRXATF-UHFFFAOYSA-N | |
78373 | |
1,1-dimethoxy-N,N-dimethylmethanamine |
C5H13NO2 | |
MFCD00008482 | |
n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane | |
CHEBI:85061 | |
CN(C)C(OC)OC |
Safety and Handling
GHS H Statement
H225-H302-H332-H315-H319-H335
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : TSCA