missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethylformamide diethyl acetal, 95%

Manufacturer:  Alfa Aesar™ L0020518

 View more versions of this product

Catalog No. AAL0020518



Specifications

Specifications

N,N-Dimethylformamide diethyl acetal
134°C to 136°C
1188-33-6
MFCD00009227
741889
BWKAYBPLDRWMCJ-UHFFFAOYSA-N
1,1-diethoxy-N,N-dimethylmethanamine
70913
95%
17°C (62°F)
50g
C7H17NO2
UN1993
1,1-diethoxy trimethyl amine
CCOC(N(C)C)OCC
147.218
147.22

Chemical Identifiers

1188-33-6
147.218
BWKAYBPLDRWMCJ-UHFFFAOYSA-N
70913
CCOC(N(C)C)OCC
C7H17NO2
MFCD00009227
1,1-diethoxy trimethyl amine
1,1-diethoxy-N,N-dimethylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

Documents