Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethylformamide dibenzyl acetal, tech. 80%

$125.51 - $1,853.31

Chemical Identifiers

Chemical Name or Material N,N-Dimethylformamide dibenzyl acetal
CAS 2016-04-8
Purity Grade Technical Grade
Molecular Formula (CH3)2NCH(OCH2C6H5)2
Formula Weight 271.36
MDL Number MFCD00014869
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1128109

  SDS
alfa aesar™
A1128109
10g Each for $125.51

AAA1128130

  SDS
alfa aesar™
A1128130
250g Each for $1,853.31

AAA1128118

  SDS
alfa aesar™
A1128118
50g Each for $495.73
Description & Specifications

Specifications

Formula Weight 271.36
Chemical Name or Material N,N-Dimethylformamide dibenzyl acetal
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 138° to 140°C (0.5mmHg)
Health Hazard 1 Warning
Molecular Formula (CH3)2NCH(OCH2C6H5)2
MDL Number MFCD00014869
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 85%
Quantity 10g
CAS 2016-04-8
Purity Grade Technical Grade

Applications
N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.

Solubility
Soluble in variety of inert solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.