missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylformamide dibenzyl acetal, tech. 80%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1128118

 View more versions of this product

Catalog No. AAA1128118



Description

Description

N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.

Solubility
Soluble in variety of inert solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

N,N-Dimethylformamide dibenzyl acetal
1.045
C17H22NO2
50 g
n,n-dimethylformamide dibenzyl acetal, n,n-dimethylformamide dibenzylacetal, 1,1-bis benzyloxy trimethylamine, methanamine, n,n-dimethyl-1,1-bis phenylmethoxy, bis benzyloxy methyl dimethylamine, acmc-1cro2, 1,1-dibenzyloxytrimethylamine, dimethyl formamide dibenzylacetal, bis benzyloxy methyldimethylamine, dimethyl formamide dibenzyl acetal
JFIKHFNGAURIIB-UHFFFAOYSA-O
N,N-dimethyl-1,1-bis(phenylmethoxy)methanamine
74837
85%
2016-04-8
138°C to 140°C (0.5 mmHg)
MFCD00014869
Moisture sensitive
Soluble in variety of inert solvents.
CN(C)C(OCC1=CC=CC=C1)OCC2=CC=CC=C2
272.37
271.36
Technical

Chemical Identifiers

2016-04-8
272.37
JFIKHFNGAURIIB-UHFFFAOYSA-O
74837
CN(C)C(OCC1=CC=CC=C1)OCC2=CC=CC=C2
C17H22NO2
MFCD00014869
n,n-dimethylformamide dibenzyl acetal, n,n-dimethylformamide dibenzylacetal, 1,1-bis benzyloxy trimethylamine, methanamine, n,n-dimethyl-1,1-bis phenylmethoxy, bis benzyloxy methyl dimethylamine, acmc-1cro2, 1,1-dibenzyloxytrimethylamine, dimethyl formamide dibenzylacetal, bis benzyloxy methyldimethylamine, dimethyl formamide dibenzyl acetal
N,N-dimethyl-1,1-bis(phenylmethoxy)methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-946-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only