Please login to your online account to display your discounted pricing

N,N-Dimethylformamide-d7, for NMR, 99.5% atom D, ACROS Organics™

$656.07 - $656.07

Chemical Identifiers

Chemical Name or Material N, N-Dimethylformamide-d7
Name Note 99.5 atom % D
CAS 4472-41-7
Purity Grade For NMR
Molecular Formula C3D7NO
Linear Formula DCON(CD3)2
Formula Weight 80.15
MDL Number MFCD00003286
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC183600100

  SDS
thermo fisher scientific
183600100
Glass bottle 10mL Each for $656.07
Description & Specifications

Specifications

Purity Grade For NMR
Physical Form Liquid
Formula Weight 80.15
Quantity 10mL
Color Clear colorless
EINECS Number 224-745-8
Density 1.0300g/mL
MDL Number MFCD00003286
Health Hazard 1 GHS Signal Word: Danger
Water 0.1% max. (Coulometric)
Deuterium (D) 99.4% min.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula DCON(CD3)2
Name Note 99.5 atom % D
Chemical Name or Material N, N-Dimethylformamide-d7
Molecular Formula C3D7NO
CAS 4472-41-7

For NMR

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapor.