Please login to your online account to display your discounted pricing

N,N-Dimethylformamide (Certified ACS), Fisher Chemical


Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

Quantity 500mL
Packaging Glass Bottle

Manufacturer: fisher scientific company  BPD119500

 View more versions of this product

Catalog No. D119-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.8%
Boiling Point 153°C
Formula Weight 73.09
Melting Point −61°C
Quantity 500mL
DOT Information DOT Class 3, : Flammable Liquid
Flash Point 58°C (136.4°F)
Physical Form Liquid
Purity Grade Certified ACS Reagent Grade
pH 6 to 8
Color Colorless
Density 0.945g/cm³
Vapor Pressure 4.9mbar at 20°C
Viscosity 0.8 mPa/s at 20°C
Decomposition Information >350°C
Alpha Vector Dimethylformamide
UN Number 2265
Packaging Glass Bottle
Water 0.15% max.
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. May cause methemoglobinemia. Irritating to eyes and skin. Lachrymator (substance which increases the flow of tears). May cause central nervous system effects. May cause adverse liver effects. May cause adverse kidney effects. May cause harm to the unborn child. Use personal protective equipment. Keep away from open flames
Cleanup Symbol Solvent
ChemAlert Storage Symbol Red
Additional Information Vapor Pressure: 4.9mbar at 20°C
MDL Number MFCD00003284
Titratable Base 0.003mEq/g max.
Residue after Evaporation 0.005% max.
Identification Pass Test
Synonym DMF
Titratable Acid 0.0005mEq/g max.
Assay Percent Range ≥99.8 %
Chemical Name or Material N, N-Dimethylformamide
CAS 68-12-2
Molecular Formula C3H7NO

Specifications

Chemical Name or Material N, N-Dimethylformamide
CAS 68-12-2
Purity Grade Certified ACS Reagent Grade
Physical Form Liquid
Assay ≥99.8%
Assay Percent Range ≥99.8 %
Molecular Formula C3H7NO
Formula Weight 73.09
MDL Number MFCD00003284
Synonym DMF
Accessories
 

Emergency Overview
Flammable liquid and vapor. May cause methemoglobinemia. Irritating to eyes and skin. Lachrymator (substance which increases the flow of tears). May cause central nervous system effects. May cause adverse liver effects. May cause adverse kidney effects. May cause harm to the unborn child. Use personal protective equipment. Keep away from open flames

WARNING!

 

SDS & Safety Documents