Please login to your online account to display your discounted pricing

N,N-Dimethylformamide (Certified ACS), Fisher Chemical


Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

Quantity 4L
Packaging Aluminum Can

Manufacturer: fisher scientific company  BPD119S4

 View more versions of this product

Catalog No. D119S-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.8%
Melting Point −61°C
Boiling Point 153°C
Formula Weight 73.09
Quantity 4L
DOT Information DOT Class 3, : Flammable Liquid
Flash Point 58°C (136.4°F)
Physical Form Liquid
Color Colorless
Purity Grade Certified ACS Reagent Grade
pH 6 to 8
Density 0.945g/cm³
Viscosity 0.8 mPa/s at 20°C
Decomposition Information >350°C
Alpha Vector Dimethylformamide
Vapor Pressure 4.9mbar at 20°C
UN Number 2265
Residue after Evaporation 0.005% max.
Titratable Base 0.003mEq/g max.
Synonym DMF
Identification Pass Test
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. May cause methemoglobinemia. Irritating to eyes and skin. Lachrymator (substance which increases the flow of tears). May cause central nervous system effects. May cause adverse liver effects. May cause adverse kidney effects. May cause harm to the unborn child. Use personal protective equipment. Keep away from open flames
Health Hazard 2 WARNING!
MDL Number MFCD00003284
ChemAlert Storage Symbol Red
Additional Information Vapor Pressure: 4.9mbar at 20°C
Packaging Aluminum Can
Assay Percent Range ≥99.8 %
Water 0.15% max.
Titratable Acid 0.0005mEq/g max.
Cleanup Symbol Solvent
CAS 68-12-2
Molecular Formula C3H7NO
Chemical Name or Material N, N-Dimethylformamide

Specifications

Chemical Name or Material N, N-Dimethylformamide
CAS 68-12-2
Purity Grade Certified ACS Reagent Grade
Physical Form Liquid
Assay ≥99.8%
Assay Percent Range ≥99.8 %
Molecular Formula C3H7NO
Formula Weight 73.09
MDL Number MFCD00003284
Synonym DMF
Accessories
 

Emergency Overview
Flammable liquid and vapor. May cause methemoglobinemia. Irritating to eyes and skin. Lachrymator (substance which increases the flow of tears). May cause central nervous system effects. May cause adverse liver effects. May cause adverse kidney effects. May cause harm to the unborn child. Use personal protective equipment. Keep away from open flames

WARNING!

 

SDS & Safety Documents