missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Dimethylformamide, B&J Brand™, for HPLC, GC, spectrophotometry, >99.9%, Honeywell Burdick & Jackson
Manufacturer: Honeywell-Burdick and Jackson CS076200
Specifications
N,N-Dimethylformamide | |
0.005 AU max. at 350 nm | |
100.0000% | |
UN2265 | |
dimethyl formamide, dimethylformamid, dimethylformamide, dimetilformamide, dmf, dmfa, dwumetyloformamid, formamide, n,n-dimethyl, n,n-dimethylmethanamide, n-formyldimethylamine | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.10 | |
HPLC | |
Returnable Steel Drum | |
0.01% max. | |
153°C |
B&J Brand™, for HPLC, GC, spectrophotometry, >99.9% | |
68-12-2 | |
C3H7NO | |
605365 | |
Miscible in Water at 20°C | |
CN(C)C=O | |
73.095 | |
CHEBI:17741 | |
>99.9% | |
0.949 g/cm3 | |
Miscible in Water at 20°C | |
58°C | |
200L |
Chemical Identifiers
68-12-2 | |
73.095 | |
dimethyl formamide, dimethylformamid, dimethylformamide, dimetilformamide, dmf, dmfa, dwumetyloformamid, formamide, n,n-dimethyl, n,n-dimethylmethanamide, n-formyldimethylamine | |
CHEBI:17741 | |
CN(C)C=O |
C3H7NO | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: N,N-Dimethylformamide