missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylformamide, 99+%, extra pure, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 116220250

 View more versions of this product

Catalog No. AC116220250



Description

Description

Specifications

Specifications

N,N-Dimethylformamide
68-12-2
100.0
99+%
HCON(CH3)2
04, 58
15, 3267
Solubility in water: soluble. Other solubilities: miscible with most organic solvents
CN(C)C=O
73.10
CHEBI:17741
Liquid
Extra Pure
Plastic drum
0.005% max.
0.945
0.2% max. (K.F.)
0.8 mPa.s (20°C)
153.0°C
-61.0°C
99%
99.0
Vapor Pressure: 4.9mbar at 20°C
C3H7NO
MFCD00003284
01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
dimethyl formamide, dimethylformamid, dimethylformamide, dimetilformamide, dmf, dmfa, dwumetyloformamid, formamide, n,n-dimethyl, n,n-dimethylmethanamide, n-formyldimethylamine
ZMXDDKWLCZADIW-UHFFFAOYSA-N
N,N-dimethylformamide
6228
73.09
≥99%
0.9450g/mL
1.4290 to 1.4320
Solubility in water: soluble. Other solubilities: miscible with most organic solvents
0.94
58°C
>350°C
Undesignated
25L

Chemical Identifiers

68-12-2
73.10
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
N,N-dimethylformamide
C3H7NO
MFCD00003284
dimethyl formamide, dimethylformamid, dimethylformamide, dimetilformamide, dmf, dmfa, dwumetyloformamid, formamide, n,n-dimethyl, n,n-dimethylmethanamide, n-formyldimethylamine
CHEBI:17741
CN(C)C=O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 200-679-5

RTECSNumber : LQ2100000

TSCA : TSCA

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only