missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylformamide, 99.8+%, for spectroscopy ACS, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 408325000

 View more versions of this product

Catalog No. AC408325000


 

Specifications Description & Specifications

Specifications

≥99.80%
ACS Grade
500mL
EcoSafPak™ Glass Bottle
0.9450g/mL
04, 58
01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
0.01 max. at 340nm
N, N-Dimethylformamide
C3H7NO
68-12-2
99.8
Vapor Pressure: 4.9mbar at 20°C
DMF
Liquid
73.09
Clear
>350°C
0.15% Max. (coulometric)
15, 3267
100.0
0.005% max.
99.8+%
HCON(CH3)2
MFCD00003284
0.003 meq/g max.
0.0005 meq/g max.
99.8%

Chemical Identifiers

68-12-2
100.0
C3H7NO
73.09
DMF
≥99.80%
99.8
99.8+%
HCON(CH3)2
MFCD00003284
N, N-Dimethylformamide, 99.8%
ACS Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 200-679-5

RTECSNumber : LQ2100000

TSCA : TSCA