Learn More
N,N-Dimethylformamide, 99.8%, for spectroscopy, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 167795000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopySpecifications
N,N-Dimethylformamide | |
0.001 meq/g max. | |
0.001 meq/g max. | |
(1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 max. at 340nm, 0.05 max. at 310nm, 0.1 max. at 295nm, 0.3 max. at 275nm, 1 max. at 270nm | |
-61.0°C | |
153.0°C | |
99.75% min. (GC) | |
C3H7NO | |
HCON(CH3)2 | |
500 mL | |
0.0015% max. | |
0.945 | |
0.94 | |
Solubility in water: soluble. Other solubilities: miscible with most organic solvents | |
CN(C)C=O | |
73.10 | |
0.8 mPa.s (20°C) | |
73.09 | |
Spectroscopy |
99+% | |
68-12-2 | |
99.75 | |
100.0 | |
0.9450g/mL | |
58°C | |
Glass bottle | |
1.4290 to 1.4320 | |
MFCD00003284 | |
04, 58 | |
01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144 | |
15, 3267 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
CHEBI:17741 | |
99.8% | |
Liquid |
Chemical Identifiers
68-12-2 | |
73.10 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
C3H7NO | |
MFCD00003284 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CHEBI:17741 | |
CN(C)C=O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wea
GHS Signal Word: Danger
EINECSNumber : 200-679-5
RTECSNumber : LQ2100000
TSCA : TSCA
RUO – Research Use Only