Please login to your online account to display your discounted pricing

N,N-Dimethylformamide, 99.8% min., ACS, EMD Millipore

$395.60 - $5,876.54

Chemical Identifiers

Chemical Name or Material N, N-Dimethylformamide
Name Note For ACS analysis
CAS 68-12-2
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula HCON(CH3)2
Formula Weight 73.09
Synonym Formic Acid Dimethylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MDX17301

 
emd millipore corporation
DX17301
4L Each for $395.60

MDX173020

 
emd millipore corporation
DX173020
200L Each for $5,876.54
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 73.09
Color Clear colorless
Quantity 4L
Packaging Glass Bottle
CAS 68-12-2
Chemical Name or Material N, N-Dimethylformamide
Name Note For ACS analysis
Assay Percent Range 99.8% min.
Molecular Formula HCON(CH3)2
Synonym Formic Acid Dimethylamide