Learn More
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
$100.31 - $348.78
Chemical Identifiers
CAS | 68-12-2 |
---|---|
Molecular Formula | C3H7NO |
Molecular Weight (g/mol) | 73.10 |
MDL Number | MFCD00003284 |
InChI Key | ZMXDDKWLCZADIW-UHFFFAOYSA-N |
Synonym | dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa |
PubChem CID | 6228 |
ChEBI | CHEBI:17741 |
IUPAC Name | N,N-dimethylformamide |
SMILES | CN(C)C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC423640250
|
Thermo Scientific Chemicals
423640250 |
25 mL | Glass Bottle |
N/A
|
|
||||
AC423645000
|
Thermo Scientific Chemicals
423645000 |
500 mL | Glass Bottle |
N/A
|
|
||||
AC423640010
|
Thermo Scientific Chemicals
423640010 |
1 L | EcoSafPak™ Glass Bottle |
N/A
|
|
||||
AC423640025
|
Thermo Scientific Chemicals
423640025 |
2.5 L | Glass Bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68-12-2 | |
73.10 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
C3H7NO | |
MFCD00003284 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CHEBI:17741 | |
CN(C)C=O |
Specifications
-61.0°C | |
99.8 | |
>350°C | |
153.0°C | |
58°C | |
Liquid | |
0.005% max. | |
0.945 | |
C3H7NO | |
HCON(CH3)2 | |
Vapor Pressure: 4.9mbar at 20°C | |
0.15% Max. (coulometric) | |
01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.09 | |
ACS Reagent |
68-12-2 | |
100.0 | |
0.9450g/mL | |
25 mL | |
Glass Bottle | |
1.4290 to 1.4320 | |
99.8+% | |
0.0005 meq/g max. | |
0.003 meq/g max. | |
MFCD00003284 | |
0.8 mPa.s (20°C) | |
04, 58 | |
15, 3267 | |
Solubility in water: soluble. Other solubilities: miscible with most organic solvents | |
CN(C)C=O | |
73.10 | |
CHEBI:17741 | |
≥99.8% | |
N,N-Dimethylformamide |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only